Merck
Mesenchymal stem cell-laden anti-inflammatory hydrogel enhances diabetic wound healing.
Scientific reports (2015-12-09)
Shixuan Chen, Junbin Shi, Min Zhang, Yinghua Chen, Xueer Wang, Lei Zhang, Zhihui Tian, Yuan Yan, Qinglin Li, Wen Zhong, Malcolm Xing, Lu Zhang, Lin Zhang
ZUSAMMENFASSUNG
The purpose of this study was to permit bone marrow mesenchymal stem cells (BMSCs) to reach their full potential in the treatment of chronic wounds. A biocompatible multifunctional crosslinker based temperature sensitive hydrogel was developed to deliver BMSCs, which improve the chronic inflammation microenvironments of wounds. A detailed in vitro investigation found that the hydrogel is suitable for BMSC encapsulation and can promote BMSC secretion of TGF-β1 and bFGF. In vivo, full-thickness skin defects were made on the backs of db/db mice to mimic diabetic ulcers. It was revealed that the hydrogel can inhibit pro-inflammatory M1 macrophage expression. After hydrogel association with BMSCs treated the wound, significantly greater wound contraction was observed in the hydrogel + BMSCs group. Histology and immunohistochemistry results confirmed that this treatment contributed to the rapid healing of diabetic skin wounds by promoting granulation tissue formation, angiogenesis, extracellular matrix secretion, wound contraction, and re-epithelialization. These results show that a hydrogel laden with BMSCs may be a promising therapeutic strategy for the management of diabetic ulcers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumpersulfat, ACS reagent, ≥98.0%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammoniumpersulfat, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammoniumpersulfat, reagent grade, 98%
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
N-Isopropylacrylamid, 97%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
N-Isopropylacrylamid, ≥99%
Sigma-Aldrich
Ammoniumpersulfat, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
Ammoniumpersulfat, BioXtra, ≥98.0%
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Ammoniumpersulfat, free-flowing, Redi-Dri, reagent grade, 98%