Merck
Oral immunostimulation of the oyster Ostrea edulis: Impacts on the parasite Bonamia ostreae.
Fish & shellfish immunology (2015-02-06)
M Prado-Alvarez, S A Lynch, A Kane, G Darmody, B G Pardo, P Martínez, J Cotterill, T Wontner-Smith, S C Culloty
ZUSAMMENFASSUNG
Bioactive compounds were orally administered to the native European oyster Ostrea edulis to evaluate the immune response and the progression of infection of the protozoan parasite Bonamia ostreae. The immunostimulants lipopolysaccharide and zymosan directly administrated to the water column induced an increase in lysozyme activity and the percentage of granulocytes in naïve oysters over a period of 7 days. In another set of experiments, zymosan and curdlan were microencapsulated in alginate and also administered to the water column to naïve and B. ostreae infected O. edulis. Oyster mortality, prevalence and intensity of infection and several immune parameters were evaluated up to 28 days post-administration. Lysozyme activity, nitric oxide production and the expression of galectin, lysozyme and superoxide dismutase increased after 24 h in both infected and uninfected oysters. Zymosan immunostimulated oysters displayed a decrease in the prevalence of B. ostreae infection not attributed to mortalities but which could be associated to the enhanced ability of immunostimulants to evoke an enhanced immune response in the oysters and reduce infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Sulfanilamid, ≥98%
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Sulfanilamid, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
p-Xylen-bis(N-Pyridiniumbromid), ≥95% (TLC)
Sigma-Aldrich
Phosphorsäure-16O4 -Lösung, 70 wt. % in D2O, 99.9 atom % 16O