Merck
Evaluating Common Humoral Responses against Fungal Infections with Yeast Protein Microarrays.
Journal of proteome research (2015-08-11)
Paulo S R Coelho, Hogune Im, Karl V Clemons, Michael P Snyder, David A Stevens
ZUSAMMENFASSUNG
We profiled the global immunoglobulin response against fungal infection by using yeast protein microarrays. Groups of CD-1 mice were infected systemically with human fungal pathogens (Coccidioides posadasii, Candida albicans, or Paracoccidioides brasiliensis) or inoculated with PBS as a control. Another group was inoculated with heat-killed yeast (HKY) of Saccharomyces cerevisiae. After 30 days, serum from mice in the groups were collected and used to probe S. cerevisiae protein microarrays containing 4800 full-length glutathione S-transferase (GST)-fusion proteins. Antimouse IgG conjugated with Alexafluor 555 and anti-GST antibody conjugated with Alexafluor 647 were used to detect antibody-antigen interactions and the presence of GST-fusion proteins, respectively. Serum after infection with C. albicans reacted with 121 proteins: C. posadasii, 81; P. brasiliensis, 67; and after HKY, 63 proteins on the yeast protein microarray, respectively. We identified a set of 16 antigenic proteins that were shared across the three fungal pathogens. These include retrotransposon capsid proteins, heat shock proteins, and mitochondrial proteins. Five of these proteins were identified in our previous study of fungal cell wall by mass spectrometry (Ann. N. Y. Acad. Sci. 2012, 1273, 44-51). The results obtained give a comprehensive view of the immunological responses to fungal infections at the proteomic level. They also offer insight into immunoreactive protein commonality among several fungal pathogens and provide a basis for a panfungal vaccine.