Merck
  • Interaction of Mycobacterium leprae with the HaCaT human keratinocyte cell line: new frontiers in the cellular immunology of leprosy.
Interaction of Mycobacterium leprae with the HaCaT human keratinocyte cell line: new frontiers in the cellular immunology of leprosy.
Experimental dermatology (2015-04-02)
Eloah C D Lyrio, Ivy C Campos-Souza, Luiz C D Corrêa, Guilherme C Lechuga, Maurício Verícimo, Helena C Castro, Saulo C Bourguignon, Suzana Côrte-Real, Norman Ratcliffe, Wim Declercq, Dilvani O Santos
ZUSAMMENFASSUNG
Leprosy is a chronic granulomatous disease caused by Mycobacterium leprae affecting the skin and peripheral nerves. Despite M. leprae invasion of the skin and keratinocytes importance in innate immunity, the interaction of these cells in vitro during M. leprae infection is poorly understood. Conventional and fluorescence optical microscopy, transmission electronic microscopy, flow cytometry and ELISA were used to study the in vitro interaction of M. leprae with the HaCaT human keratinocyte cell line. Keratinocytes uptake of M. leprae is described, and modulation of the surface expression of CD80 and CD209, cathelicidin expression and TNF-α and IL-1β production of human keratinocytes are compared with dendritic cells and macrophages during M. leprae interaction. This study demonstrated that M. leprae interaction with human keratinocytes enhanced expression of cathelicidin and greatly increased TNF-α production. The highest spontaneous expression of cathelicidin was by dendritic cells which are less susceptible to M. leprae infection. In contrast, keratinocytes displayed low spontaneous cathelicidin expression and were more susceptible to M. leprae infection than dendritic cells. The results show, for the first time, an active role for keratinocytes during infection by irradiated whole cells of M. leprae and the effect of vitamin D on this process. They also suggest that therapies which target cathelicidin modulation may provide novel approaches for treatment of leprosy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Osmiumtetroxid, Sealed ampule.
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Interleukin-4 human, potency:, IL-4, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Interleukin-4 from mouse, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Interleukin-4 from rat, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)