Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Hexamethyldisilazan, reagent grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
N,N′-Methylenbisacrylamid, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenbis(acrylamid), 99%
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamid, synthesis grade, ≥95%
Sigma-Aldrich
Hexamethyldisilazan, ReagentPlus®, 99.9%
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
N,N′-Methylenbisacrylamid -Lösung, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Kohlenstoff, mesoporös, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Kohlenstoff, mesoporös, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, powder, electrophoresis grade
Sigma-Aldrich
Kohlenstoff, mesoporös
Sigma-Aldrich
Kohlenstoff, mesoporös, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
N,N′-Methylenbisacrylamid, suitable for electrophoresis (after filtration or allowing insolubles to settle)