Merck
Serum antibody immunoreactivity to equine zona protein after SpayVac vaccination.
Theriogenology (2015-04-30)
Tracy A Mask, Kathryn A Schoenecker, Albert J Kane, Jason I Ransom, Jason E Bruemmer
ZUSAMMENFASSUNG
Immunocontraception with porcine ZP (pZP) can be an effective means of fertility control in feral horses. Previous studies suggest that antibodies produced after pZP vaccination may both inhibit fertilization and cause follicular dysgenesis. Zonastat-H, PZP-22, and SpayVac are three pZP vaccines proposed for use in horses. Although all these vaccines contain the pZP antigen, variations in antigen preparation and vaccine formulation lead to differences in antigenic properties among them. Likewise, despite numerous efficacy and safety studies of Zonastat-H and PZP-22, the contraceptive mechanisms of SpayVac remain unclear. The preparation of pZP for SpayVac is thought to include more nonzona proteins, making it less pure than the other two vaccines. This may result in increased antigenicity of the vaccine. We therefore investigated the immunoreactivity of serum antibodies from SpayVac-vaccinated mares to equine zona protein. Western blot analyses revealed an immunoreactivity of these antibodies to protein isolated from mature equine oocytes, ZP, follicular tissues, and ovarian tissues. Immunohistochemical analyses were used to locate the binding of serum antibodies to the ZP of immature oocytes in ovarian stromal tissue. We also found serum antibodies from SpayVac-treated mares to be predominantly specific for zona protein 3. Collectively, our results suggest a model where serum antibodies produced in response to SpayVac vaccination are immunoreactive to equine zona protein in vitro. Our study lends insight into the contraceptive mechanisms underlying the infertility observed after SpayVac vaccination.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamid, synthesis grade, ≥95%
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Bromphenolblau, titration: suitable
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri