Merck
  • Synthesis and physicochemical characterization of the impurities of pemetrexed disodium, an anticancer drug.
Synthesis and physicochemical characterization of the impurities of pemetrexed disodium, an anticancer drug.
Molecules (Basel, Switzerland) (2015-06-04)
Olga Michalak, Mariusz M Gruza, Anna Witkowska, Iwona Bujak, Piotr Cmoch
ZUSAMMENFASSUNG
A physicochemical characterization of the process-related impurities associated with the synthesis of pemetrexed disodium was performed. The possibility of pemetrexed impurities forming has been mentioned in literature, but no study on their structure has been published yet. This paper describes the development of the synthesis methods for these compounds and discusses their structure elucidation on the basis of two-dimensional NMR experiments and MS data. The identification of these impurities should be useful for the quality control during the production of the pemetrexed disodium salt.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Kaliumphosphat, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Kaliumphosphat, reagent grade, ≥98.0%
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Kaliumphosphat -Lösung, 1.0 M
Sigma-Aldrich
Aluminiumoxid, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminiumoxid, 99.997% trace metals basis
Sigma-Aldrich
Aluminiumoxid, single crystal substrate, <0001>
Sigma-Aldrich
Kaliumphosphat, 99.95% trace metals basis
Sigma-Aldrich
Aluminiumoxid, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminumoxid, mesoporös, MSU-X (wormhole), average pore size 3.8 nm