Merck
Ginsenoside Rg1 Ameliorates Motor Function in an Animal Model of Parkinson's Disease.
Pharmacology (2015-06-13)
Wenda Jiang, Zhejian Wang, Yiyang Jiang, Meili Lu, Xin Li
ZUSAMMENFASSUNG
Parkinson's disease (PD) is a neurodegenerative disorder characterized by loss of dopaminergic neurons in the substantia nigra (SN) and diminished dopamine levels in the striatum. Accumulating evidence supported that ginsenoside Rg1, the major pharmacologically active compound of ginseng, has a wide range of neurotrophic and neuroprotective effects under physiological and pathological conditions. Although Rg1 administration protects dopaminergic neurons in a rat model of PD, it is unclear if Rg1 treatment ameliorates motor function in PD. Using the neurotoxin 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) that causes dopaminergic neurodegeneration, we investigated the effect of Rg1 on 3 tests of motor behaviors in mice: the accelerating rotarod, wire suspension and pole tests. The results showed that Rg1 treatment (10 mg/kg, i.p.) succeeded in restoring motor functions to physiological level in MPTP-treated mice. Importantly, these behavioral ameliorations were accompanied by an attenuation of the MPTP-induced loss of dopaminergic neurons in the SN and striatum. These findings indicate that Rg1 can significantly rescue the deficit of motor function in mice model of PD, and suggest that Rg1 may be a potential therapeutic agent against PD and related disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Urethan, ≥99%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Urethan, ≥99.0% (GC)
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Sigma-Aldrich
DL-Tyrosin, 99%