- Two Schiff-base fluorescent sensors for selective sensing of aluminum (III): Experimental and computational studies.
Two Schiff-base fluorescent sensors for selective sensing of aluminum (III): Experimental and computational studies.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-08-02)
Jing-Can Qin, Xiao-Ying Cheng, Ran Fang, Ming-Fang Wang, Zheng-Yin Yang, Tian-Rong Li, Yong Li
PMID26232579
ZUSAMMENFASSUNG
Two Schiff-base fluorescent sensors have been synthesized, which both can act as fluorescent probes for Al(3+), upon addition of Al(3+), they exhibit a large fluorescence enhancement which might be attributed to the formation of 1:1 ligand-Al complexes which inhibit photoinduced electron transfer (PET) progress, and that the proposed binding modes of the sensors and Al(3+) are identified by theoretical calculations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)