Merck
HNA antibody-mediated neutrophil aggregation is dependent on serine protease activity.
Vox sanguinis (2015-06-19)
T Berthold, N Schubert, S Muschter, M Rohr, J Wesche, A Reil, J Bux, T Bakchoul, A Greinacher
ZUSAMMENFASSUNG
Transfusion-related acute lung injury (TRALI) is often caused by antibodies against human neutrophil alloantigen-2 (HNA-2) and HNA-3a. Neutrophil aggregation is considered as a major cause of TRALI, but little is known about how HNA antibodies initiate this process. We explored mechanisms involved in neutrophil aggregation induced by HNA-2 and HNA-3a antibodies. Isolated neutrophils were pretreated with broad-spectrum or specific inhibitors against different cell functions or proteases. Granulocyte agglutination test (GAT) was performed with serially diluted anti-HNA-2 and anti-HNA-3a plasmas or control plasma, and reactivity was evaluated microscopically. Reactive oxygen species (ROS) production in neutrophils was investigated using a lucigenin-based chemiluminescence assay. HNA-2 and HNA-3a antibody-mediated neutrophil aggregation was inhibited by pretreatment with formaldehyde, iodoacetamide and the serine protease inhibitors Pefabloc-SC, N-p-tosyl-L-phenylalanine chloromethyl ketone (TPCK) and Nα-tosyl-L-lysine chloromethyl ketone hydrochloride (TLCK). In contrast, inhibition of actin polymerization, respiratory burst, cysteine proteases, metalloproteases or aspartic proteases did not affect neutrophil aggregation. Furthermore, HNA-3a antibodies did not directly cause ROS production in neutrophils. Aggregation of neutrophils induced by HNA-2 and HNA-3a antibodies is an active process and depends on trypsin- or chymotrypsin-like serine proteases but is not dependent on the production of ROS. These findings may open new prospects for the pharmacologic prevention of neutrophil-associated acute lung injury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Supelco
D-(+)-Glukose, analytical standard
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Diphenyleniodoniumchlorid, ≥98%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
DL-Cystein, technical grade
Sigma-Aldrich
D-(+)-Glukose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
DL-Serin, ≥98% (TLC)
Sigma-Aldrich
D-(+)-Glukose, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
DL-Serin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)