Merck
  • Simultaneous determination of arbutin and its decomposed product hydroquinone in whitening creams using high-performance liquid chromatography with photodiode array detection: Effect of temperature and pH on decomposition.
Simultaneous determination of arbutin and its decomposed product hydroquinone in whitening creams using high-performance liquid chromatography with photodiode array detection: Effect of temperature and pH on decomposition.
International journal of cosmetic science (2015-04-11)
J S Jeon, B H Kim, S H Lee, H J Kwon, H J Bae, S K Kim, J A Park, J H Shim, A M Abd El-Aty, H C Shin
ZUSAMMENFASSUNG
Arbutin is an effective agent for the treatment of melanin disorders. Arbutin may be converted to hydroquinone under conditions of high temperature, ultraviolet (UV) radiation and dilute acid. The aim of the current study was to develop an analytical method to determine the levels of arbutin and hydroquinone in whitening cosmetic products using high-performance liquid chromatography with photodiode array detection (HPLC-DAD). In addition, we investigated the effects of high temperature and pH on the decomposition of arbutin. Samples extracted using two-step sonications were separated on a C18 column using a gradient mobile phase consisting of water and methanol. A 60-mm (40 μL) DAD cell was used to enhance the sensitivity of hydroquinone determination. Thermal decomposition of arbutin was evaluated at temperatures ranging from 60 to 120°C for 1-36 h. The method showed good linearity (R(2) ≥ 0.9997), precision (relative standard deviation, RSD < 5%) and acceptable extraction recovery (90-102.6%). The limits of quantitation for arbutin and hydroquinone were 0.0085 and 0.0119 μg mL(-1) , respectively. One sample of 21 cosmetic products tested contained arbutin at a concentration 1.61 g 100 g(-1) cream and 0.12 g 100 g(-1) cream of hydroquinone. Arbutin (327.18 ppm) decomposed after 6 h at 120°C and produced 10.73 ppm of hydroquinone. The developed method is simple to detect both arbutin and hydroquinone simultaneously in cosmetic products, at an adequate level of sensitivity. Notably, temperature and pH did not influence the decomposition of arbutin to hydroquinone in a 2% arbutin cream.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Hydrochinon, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochinon, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Arbutin, ≥98% (HPLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochinon, meets USP testing specifications
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.