Merck
  • Morphological and molecular characterization of a sexually reproducing colony of the booklouse Liposcelis bostrychophila (Psocodea: Liposcelididae) found in Arizona.
Morphological and molecular characterization of a sexually reproducing colony of the booklouse Liposcelis bostrychophila (Psocodea: Liposcelididae) found in Arizona.
Scientific reports (2015-05-28)
Qianqian Yang, Zuzana Kučerová, Steve J Perlman, George P Opit, Edward L Mockford, Adi Behar, Wyatt E Robinson, Václav Stejskal, Zhihong Li, Renfu Shao
ZUSAMMENFASSUNG
The booklouse, Liposcelis bostrychophila, is a worldwide pest of stored products. For decades, only thelytokous parthenogenetic reproduction was documented in L. bostrychophila. Male L. bostrychophila were first found in Hawaii in 2002. In 2009, a sexual strain was found in Arizona. We examined the morphology of both males and females of the Arizona strain and compared the Arizona sexual strain with the Hawaii sexual strain and the parthenogenetic strains of L. bostrychophila. The sexual and parthenogenetic strains show some differences in eye morphology. To examine the relationship between sexual and asexual lineages, we sequenced the mitochondrial 12S and 16S ribosomal RNA genes of males and females from the Arizona strain. Phylogenetic analyses of L. bostrychophila individuals revealed that: 1) the sexually reproducing colony found in Arizona contains two closely related mitochondrial DNA haplotypes--one present in only females and the other in both males and females; and 2) the Arizona sexual strain was most closely related to a parthenogenetic strain in Illinois. We detected Rickettsia in all of the parthenogenetic individuals we checked but not in any Arizona sexual individuals. Further evidence is required to establish whether the presence of Rickettsia is linked to asexual reproduction in Liposcelis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)