Merck
  • Assembly of IMPDH2-based, CTPS-based, and mixed rod/ring structures is dependent on cell type and conditions of induction.
Assembly of IMPDH2-based, CTPS-based, and mixed rod/ring structures is dependent on cell type and conditions of induction.
Journal of genetics and genomics = Yi chuan xue bao (2015-07-15)
Gerson Dierley Keppeke, S John Calise, Edward K L Chan, Luis Eduardo C Andrade
ZUSAMMENFASSUNG
Inhibition of guanosine triphosphate (GTP) and cytidine triphosphate (CTP) biosynthetic pathways induces cells to assemble rod/ring (RR) structures, also named cytoophidia, which consist of the enzymes cytidine triphosphate synthase (CTPS) and inosine-5'-monophosphate dehydrogenase 2 (IMPDH2). We aim to explore the interaction of CTPS and IMPDH2 in the generation of RR structures. HeLa and COS-7 cells were cultured in normal conditions or in the presence of 6-diazo-5-oxo-L-norleucine (DON), ribavirin, or mycophenolic acid (MPA). Over 90% of DON-treated cells presented RR structures. In HeLa cells, 35% of the RR structures were positive for IMPDH2 alone, 26% were CTPS alone, and 31% were IMPDH2/CTPS mixed, while in COS-7 cells, 42% of RR were IMPDH2 alone, 41% were CTPS alone, and 10% were IMPDH2/CTPS mixed. Ribavirin and MPA treatments induced only IMPDH2-based RR. Cells were also transfected with an N-terminal hemagglutinin (NHA)-tagged CTPS1 construct. Over 95% of NHA-CTPS1 transfected cells with DON treatment presented IMPDH2-based RR and almost 100% presented CTPS1-based RR; when treated with ribavirin, over 94% of transfected cells presented IMPDH2-based RR and 37% presented CTPS1-based RR, whereas 2% of untreated transfected cells presented IMPDH2-based RR and 28% presented CTPS1-based RR. These results may help in understanding the relationship between CTP and GTP biosynthetic pathways, especially concerning the formation of filamentous RR structures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Methoxy-2-propyl-acetat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
Guanosin, ≥98%
Sigma-Aldrich
6-Diazo-5-oxo-L-Norleucin, powder
Sigma-Aldrich
Mycophenolsäure, ≥98%
Sigma-Aldrich
Guanosin, BioReagent, suitable for cell culture
Sigma-Aldrich
Ribavirin, antiviral
Sigma-Aldrich
Mecamylamin -hydrochlorid
Sigma-Aldrich
Mycophenolsäure, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanosin, ≥97.0% (HPLC)