Merck
Storage Time and Temperature Effects on Histamine Production in Tuna Salad Preparations.
Journal of food protection (2015-07-22)
Susan McCarthy, Kristin Bjornsdottir-Butler, Ronald Benner
ZUSAMMENFASSUNG
Scombrotoxin fish poisoning (SFP), also known as histamine (Hst) poisoning, has been associated with consumption of scombroid-type fish, including tuna and tuna fish products. Preparation of commercial tuna salad contaminated with Hstproducing bacteria (HPB), combined with time-temperature abuse, can present a food safety hazard. A potential source of HPB is raw ingredients, such as celery and onions. The objectives of this study were to determine whether raw ingredients can be a source of HPB and to ascertain the effects of storage time (up to 4 days or 4 weeks) and temperature (4, 10, 18, 25, 30°C) on growth and Hst production by high-HPB (>1,000 ppm of Hst) in tuna salad preparations. Pantoea-Erwinia, Erwinia persicinus, Erwinia spp., and Enterobacter pyrinus isolated from celery in this study were used to inoculate tuna salad and tuna salad with celery or onion. HPB numbers were 0.7 to 4.3 log most probable number per g higher in the presence of celery or onion versus plain tuna salad (3:1 tuna:mayonnaise). E. pyrinus-inoculated plain tuna salad and tuna salad with celery and onion had >500 ppm of Hst after 2 days at 30°C and 4 days at 25°C. E. pyrinus-inoculated salad with celery and onion had >500 ppm of Hst after 4 days at 18°C and 2 weeks at 10°C. Raw celery can introduce HPB into tuna salad, which can cause SFP if the product is time-temperature abused. Tuna salad products must be refrigerated at ≤4°C to prevent growth and Hst production by the HPB used in this study, to protect consumers from potential SFP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phthaldialdehyd, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Histamin -dihydrochlorid, ≥99% (TLC), powder
Sigma-Aldrich
Phthaldialdehyd, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Phthaldialdehyd, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Histamin -dihydrochlorid, ≥99.0% (AT)
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O