Merck
Few-layered MoSe2 nanosheets as an advanced electrode material for supercapacitors.
Dalton transactions (Cambridge, England : 2003) (2015-08-05)
Suresh Kannan Balasingam, Jae Sung Lee, Yongseok Jun
ZUSAMMENFASSUNG
We report the synthesis of few-layered MoSe2 nanosheets using a facile hydrothermal method and their electrochemical charge storage behavior. A systematic study of the structure and morphology of the as-synthesized MoSe2 nanosheets was performed. The downward peak shift in the Raman spectrum and the high-resolution transmission electron microscopy images confirmed the formation of few-layered nanosheets. The electrochemical energy-storage behavior of MoSe2 nanosheets was also investigated for supercapacitor applications in a symmetric cell configuration. The MoSe2 nanosheet electrode exhibited a maximum specific capacitance of 198.9 F g(-1) and the symmetric device showed 49.7 F g(-1) at a scan rate of 2 mV s(-1). A capacitance retention of approximately 75% was observed even after 10 000 cycles at a high charge-discharge current density of 5 A g(-1). The two-dimensional MoSe2 nanosheets exhibited a high specific capacitance and good cyclic stability, which makes it a promising electrode material for supercapacitor applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumborhydrid, powder, ≥98.0%
Sigma-Aldrich
Natriumborhydrid, ReagentPlus®, 99%
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Natriumborhydrid, granular, 99.99% trace metals basis
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Natriumborhydrid, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Natriumborhydrid, granular, 10-40 mesh, 98%
Sigma-Aldrich
Natriumborhydrid -Lösung, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Natriummolybdat, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumborhydrid, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Natriumborhydrid -Lösung, 2.0 M in triethylene glycol dimethyl ether
Selen, foil, 25x25mm, thickness 3mm, 99.95%
Selen, pellets, < 5mm, ≥99.999%