Merck
  • Novel cerium doped glass-reinforced hydroxyapatite with antibacterial and osteoconductive properties for bone tissue regeneration.
Novel cerium doped glass-reinforced hydroxyapatite with antibacterial and osteoconductive properties for bone tissue regeneration.
Biomedical materials (Bristol, England) (2015-09-24)
D S Morais, S Fernandes, P S Gomes, M H Fernandes, P Sampaio, M P Ferraz, J D Santos, M A Lopes, N Sooraj Hussain
ZUSAMMENFASSUNG
The aim of this work was to develop a bioactive bone substitute with an effective antibacterial ability based on a cerium (Ce) doped glass-reinforced hydroxyapatite (GR-HA) composite. Developed composites were physicochemically characterized, using x-ray diffraction (XRD) analysis, SEM, energy dispersive x-ray spectroscopy (EDS), and flexural bending strength (FBS) tests. X-ray photoelectron spectroscopy (XPS) analysis was performed to analyze the oxidation state of Ce in the prepared doped glass. The antimicrobial activity of the composites was evaluated against Staphylococcus aureus, Staphylococcus epidermidis and Pseudomonas aeruginosa; whether the cytocompatibility profile was assayed with human osteoblastic-like cells (Mg-63 cell line). The results revealed that the Ce inclusion in the GR-HA matrix induced the antimicrobial ability of the composite. In addition, Ce-doped materials reported an adequate biological behavior following seeding of osteoblastic populations, by inducing cell adhesion and proliferation. Developed materials were also found to enhance the expression of osteoblastic-related genes. Overall, the developed GR-HA_Ce composite is a prospective candidate to be used within the clinical scenario with a successful performance due to the effective antibacterial properties and capability of enhancing the osteoblastic cell response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Natriumcarbonat, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Natriumcarbonat, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Calciumhydroxid, ACS reagent, ≥95.0%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Natriumcarbonat, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Natriumcarbonat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorpentoxid, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Natriumcarbonat, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumcarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Calciumphosphat, 98.0-105.0%
Sigma-Aldrich
Phosphorpentoxid, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Calciumhydroxid, 99.995% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Phosphorpentoxid, desiccant, with moisture indicator
Supelco
Phosphorpentoxid, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorpentoxid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Zweibasisches Kalziumphosphat, meets USP testing specifications