Merck
Effects of acute heat stress on lipid metabolism of bovine primary adipocytes.
Journal of dairy science (2015-10-05)
M P Faylon, L H Baumgard, R P Rhoads, D M Spurlock
ZUSAMMENFASSUNG
Heat stress (HS) affects numerous physiological processes including nutrient partitioning and lipid metabolism. Objectives of this study were to evaluate how acute HS affects lipid metabolism in subcutaneous adipose tissue of dairy cattle. Adipose tissue biopsies were performed on Holstein cows for bovine primary adipocyte isolation and cultured at either 42°C (HS) or 37°C (thermal neutral, TN). Adipocytes were incubated with increasing isoproterenol (ISO), and with increasing concentrations of insulin in the presence of ISO to evaluate changes in lipolysis. Incorporation of radioactive acetate into lipids was measured as an indicator of lipogenesis. Abundance and phosphorylation of several lipolytic and lipogenic proteins were also measured. Adipocytes exposed to HS had an elevated maximal response to ISO and were more sensitive to lipolytic stimulation by ISO compared with cells cultured at TN. Thermal treatment did not affect the antilipolytic effects of insulin in the presence of ISO. Lipogenesis measured as acetate incorporation was not altered by HS, but a temperature by insulin interaction was observed for the regulation of acetyl CoA carboxylase, such that the presence of insulin resulted in a reduction in phosphorylation of acetyl CoA carboxylase in adipocytes cultured at TN but not HS conditions. Results of this study demonstrate that acute HS has a direct effect on the regulation of lipolysis and the rate-limiting enzyme of lipogenesis in isolated bovine adipocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)