Merck
  • Identification of target proteins of mangiferin in mice with acute lung injury using functionalized magnetic microspheres based on click chemistry.
Identification of target proteins of mangiferin in mice with acute lung injury using functionalized magnetic microspheres based on click chemistry.
Journal of agricultural and food chemistry (2015-10-22)
Jiajia Wang, Yan Nie, Yunjuan Li, Yuanyuan Hou, Wei Zhao, Jiagang Deng, Peng George Wang, Gang Bai
ZUSAMMENFASSUNG
Prevention of the occurrence and development of inflammation is a vital therapeutic strategy for treating acute lung injury (ALI). Increasing evidence has shown that a wealth of ingredients from natural foods and plants have potential anti-inflammatory activity. In the present study, mangiferin, a natural C-glucosyl xanthone that is primarily obtained from the peels and kernels of mango fruits and the bark of the Mangifera indica L. tree, alleviated the inflammatory responses in lipopolysaccharide (LPS)-induced ALI mice. Mangiferin-modified magnetic microspheres (MMs) were developed on the basis of click chemistry to capture the target proteins of mangiferin. Mass spectrometry and molecular docking identified 70 kDa heat-shock protein 5 (Hspa5) and tyrosine 3-monooxygenase (Ywhae) as mangiferin-binding proteins. Furthermore, an enzyme-linked immunosorbent assay (ELISA) indicated that mangiferin exerted its anti-inflammatory effect by binding Hspa5 and Ywhae to suppress downstream mitogen-activated protein kinase (MAPK) signaling pathways. Thoroughly revealing the mechanism and function of mangiferin will contribute to the development and utilization of agricultural resources from M. indica L.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eisen(II)-chlorid Tetrahydrat, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Eisen(II)-chlorid Tetrahydrat, ReagentPlus®, 98%
Sigma-Aldrich
Eisen(II)-chlorid Tetrahydrat, 99.99% trace metals basis
Sigma-Aldrich
Mangiferin Mangifera indica
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)