Merck
  • Furin-dependent CCL17-fused recombinant toxin controls HTLV-1 infection by targeting and eliminating infected CCR4-expressing cells in vitro and in vivo.
Furin-dependent CCL17-fused recombinant toxin controls HTLV-1 infection by targeting and eliminating infected CCR4-expressing cells in vitro and in vivo.
Retrovirology (2015-08-21)
Masateru Hiyoshi, Kazu Okuma, Seiji Tateyama, Kazuya Takizawa, Masumichi Saito, Madoka Kuramitsu, Kumiko Araki, Kazuhiro Morishita, Seiji Okada, Naoki Yamamoto, Arya Biragyn, Kazunari Yamaguchi, Isao Hamaguchi
ZUSAMMENFASSUNG
Adult T-cell leukemia (ATL) is caused by human T-cell leukemia virus type 1 (HTLV-1) infection. However, there are no therapies to prevent ATL development in high-risk asymptomatic carriers. To develop a therapy targeting HTLV-1-infected cells that are known to express CCR4 frequently, we tested whether truncated Pseudomonas exotoxin (PE38) fused to a CCR4 ligand, CCL17/thymus and activation-regulated chemokine (TARC), selectively eliminates such cells. Our data show that TARC-PE38 efficiently killed HTLV-1-infected cell lines. It also shrank HTLV-1-associated solid tumors in an infected-cell-engrafted mouse model. In HTLV-1-positive humanized mice, TARC-PE38 markedly inhibited the proliferation of HTLV-1-infected human CD4(+)CD25(+) or CD4(+)CD25(+)CCR4(+) cells and reduced the proviral loads (PVLs) in peripheral blood mononuclear cells (PBMCs). Importantly, TARC-PE38 significantly reduced the PVLs in PBMCs obtained from asymptomatic carriers. We show that the cytotoxicity of TARC-PE38 is mediated by the expression of the proprotein convertase, furin. The expression of furin was enhanced in HTLV-1-infected cells and correlated positively with PVLs in HTLV-1-infected individuals, suggesting that infected cells are more susceptible to TARC-PE38 than normal cells. TARC-PE38 robustly controls HTLV-1 infection by eliminating infected cells in both a CCR4- and furin-dependent manner, indicating the excellent therapeutic potential of TARC-PE38.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Humanes Interleukin-2 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Magnesium-methylcarbonat -Lösung, 2.0 M in DMF
Sigma-Aldrich
Interleukin-2, recombinant, expressed in E. coli, ~10000 U/mL