Merck
  • Rifabutin reduces systemic exposure of an antimalarial drug 97/78 upon co-administration in rats: An in-vivo &in-vitro analysis.
Rifabutin reduces systemic exposure of an antimalarial drug 97/78 upon co-administration in rats: An in-vivo &in-vitro analysis.
Asian Pacific journal of tropical medicine (2015-09-01)
Yeshwant Singh, Mahendra Kumar Hidau, Shio Kumar Singh
ZUSAMMENFASSUNG
To determine the potential drug-drug interactions between anti-malarial candidate 97/78 and anti-tubercular drug rifabutin in-vivo in rats followed by in-vitro investigation of the underlying mechanisms of drug interaction. Single oral dose study was conducted in male and female rats at 40 mg/kg and 70 mg/kg for 97/78 and rifabutin respectively. It was reported that rifabutin co-administration altered pharmacokinetics of 97/63 (active metabolite of 97/78). A significant decrease was reported in the systemic exposure of 97/63 by a factor of 3-4. The AUC0-last values were (4.03 ± 0.60) and (5.44 ± 1.15) μg h mL(-1) upon 97/78 administration alone, while the values were decreased to (1.13 ± 0.10) and (1.23 ± 1.13) μg h mL(-1) upon rifabutin co-administration in male and female rats respectively. Statistically significant differences were also reported in Cmax and Tmax values upon rifabutin co-administration. In-vitro drug metabolism study in rat liver microsomes has shown that the metabolism of 97/63 was increased by 10%-12% upon rifabutin co-incubation. The extent of plasma protein binding of 97/63 was found to be decreased from 54%-55% to 6%-8% upon rifabutin addition. It was concluded that rifabutin co-administration altered PK parameters of 97/63 in SD rats. However, no intersex influences were reported in the interaction pattern. The results obtained in the in-vivo study were well correlated with the in-vitro findings and can further be applied to explore other aspects of potential drug interactions between these two drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Methylamin -Lösung, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Methylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Methylamin -Lösung, 2.0 M in methanol
Sigma-Aldrich
1-Aminobenzotriazol
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Rifabutin, >98% (HPLC), powder