Merck
  • Polypyrrole-coated magnetic nanoparticles as an efficient adsorbent for RB19 synthetic textile dye: Removal and kinetic study.
Polypyrrole-coated magnetic nanoparticles as an efficient adsorbent for RB19 synthetic textile dye: Removal and kinetic study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-05-16)
Maryam Shanehsaz, Shahram Seidi, Yousefali Ghorbani, Seyed Mohammad Reza Shoja, Shohre Rouhani
ZUSAMMENFASSUNG
The present work deals with the first attempt to study the removal of synthetic textile dye, reactive blue 19 (RB19), using the magnetic Fe3O4 nanoparticles modified by pyrrole (PPy@Fe3O4 MNPs) as an efficient adsorbent. The nanoadsorbent was synthesized using chemical co-precipitation. Scanning electron microscopy and FT-IR were used to characterize nanoparticles. Factors affecting the dye adsorption including the pH of the dye solution, amount of adsorbent and contact time were also further investigated. Sorption of the RB19 on PPy@Fe3O4 MNPs reached to equilibrium at contact time less than 10 min and fitted well to the Langmuir adsorption model with a maximum adsorption capacity of 112.36 mg g(-1). Experiments for adsorption kinetic were carried out and the data fitted well according to a pseudo-second-order kinetic model. Moreover, the MNPs were recovered with over than 90% efficiency using methanol as elution agent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Eisen(II)-chlorid, 98%
Sigma-Aldrich
Eisen(III)-chlorid, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Eisen(III)-chlorid, ≥99.9% trace metals basis
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Eisen(II)-chlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Eisen(II)-chlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Eisen(III)-chlorid, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Eisen(II)-chlorid, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.