Merck
Toward surfactant-free and water-free microemulsions.
Journal of colloid and interface science (2015-05-20)
Veronika Fischer, Julien Marcus, Didier Touraud, Olivier Diat, Werner Kunz
ZUSAMMENFASSUNG
It was recently demonstrated that a nano-clustering was present in the monophasic "pre-Ouzo" region of ternary liquid mixtures without surfactants. The goal of this work is to check if this nano-clustering is also present in the surfactant-free and water-free "green" microemulsions glycerol/ethanol/1-octanol and deep eutectic solvent/tetrahydrofurfuryl alcohol/diethyl adipate. The deep eutectic solvents used instead of water were ethylene glycol-choline chloride (molar ratio 4-1) and urea-choline chloride (molar ratio 2-1). To our knowledge this is the first time that deep eutectic solvents were used to formulate microemulsions. The surfactant-free and water-free microemulsions were studied using phase diagrams, dynamic light scattering, and small-angle X-ray scattering. The presence of aggregate fluctuations was demonstrated and they were found to be independent of molecular critical fluctuations, except when approaching the critical point where the critical phenomenon is superimposed to the signal. These structures have similarities to classical microemulsions but, in contrast to them, without having a sharp interface between the non-miscible phases, much as it was the case for systems previously investigated like water/ethanol/oil, where the oil was 1-octanol, fragrance molecules, or mosquito repellents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Cholinchlorid, ≥98%
Sigma-Aldrich
Ethylenglycol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Cholinchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Cholinchlorid, ≥99%
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Cholinchlorid, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Tetrahydrofurfurylalkohol, 99%
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethylenglykol 5 M -Lösung
Sigma-Aldrich
Ethylenglycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Diethyladipat, ReagentPlus®, 99%
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Supelco
Ethylenglycol -Lösung, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Tetrahydrofurfurylalkohol, 98%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
Tetrahydrofurfurylalkohol, ≥98%