Merck
  • Evaluation of the factors influencing the resultant diameter of the electrospun gelatin/sodium alginate nanofibers via Box-Behnken design.
Evaluation of the factors influencing the resultant diameter of the electrospun gelatin/sodium alginate nanofibers via Box-Behnken design.
Materials science & engineering. C, Materials for biological applications (2015-10-20)
Seza Özge Gönen, Melek Erol Taygun, Sadriye Küçükbayrak
ZUSAMMENFASSUNG
This article presented a study on the effects of solution properties (i.e., gelatin concentration, alginate concentration, content of alginate solution in the blend solution, and content of acetic acid in the solvent of gelatin solution) on the average diameter of electrospun gelatin/sodium alginate nanofibers, as well as its standard deviation. For this purpose, blend solutions of two natural polymers (gelatin and sodium alginate) were prepared both in the absence and presence of ethanol. Response surface methodology based on a three-level, four-variable Box-Benkhen design was employed to define quadratic relationships between the responses and the solution properties. The individual and interactive effects of the solution properties were determined. Moreover, the adequacy of the models was verified by the validation experiments. Results showed that the average diameters of the resultant nanofibers were 68-166 nm and 90-155 nm in the absence and presence of ethanol, respectively. The experimental results were in good agreement with the predicted response values. Hence, this study provides an overview on the fabrication of gelatin/sodium alginate nanofibers with targeted diameter, which may have potential to be used in the field of tissue engineering.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C