Merck
  • Preparation and mechanical properties of photo-crosslinked poly(trimethylene carbonate) and nano-hydroxyapatite composites.
Preparation and mechanical properties of photo-crosslinked poly(trimethylene carbonate) and nano-hydroxyapatite composites.
Clinical hemorheology and microcirculation (2015-03-31)
Mike A Geven, Davide Barbieri, Huipin Yuan, Joost D de Bruijn, Dirk W Grijpma
ZUSAMMENFASSUNG
Composite materials of photo-crosslinked poly(trimethylene carbonate) and nanoscale hydroxyapatite were prepared and their mechanical characteristics for application as orbital floor implants were assessed. The composites were prepared by solvent casting poly(trimethylene carbonate) macromers with varying amounts of nano-hydroxyapatite and subsequent photo-crosslinking. The incorporation of the nano-hydroxyapatite into the composites was examined by thermogravimetric analysis, scanning electron microscopy and gel content measurements. The mechanical properties were investigated by tensile testing and trouser tearing experiments. Our results show that nano-hydroxyapatite particles can readily be incorporated into photo-crosslinked poly(trimethylene carbonate) networks. Compared to the networks without nano-hydroxyapatite, incorporation of 36.3 wt.% of the apatite resulted in an increase of the E modulus, yield strength and tensile strength from 2.2 MPa to 51 MPa, 0.5 to 1.4 N/mm2 and from 1.3 to 3.9 N/mm2, respectively. We found that composites containing 12.4 wt.% nano-hydroxyapatite had the highest values of strain at break, toughness and average tear propagation strength (376% , 777 N/mm2 and 3.1 N/mm2, respectively).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Propylencarbonat, anhydrous, 99.7%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propan, dist., ≥98.0% (GC)
Sigma-Aldrich
Calciumhydrid, reagent grade, 95% (gas-volumetric)
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propan, 97%
Sigma-Aldrich
Calciumhydrid, powder, 0-2 mm, reagent grade, ≥90% (gas-volumetric)
Sigma-Aldrich
Calciumhydrid, 99.9% trace metals basis
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Calciumhydrid, purum p.a., ≥97.0% (gas-volumetric), powder
Sigma-Aldrich
Calciumhydrid, powder, 99.99% trace metals basis