Merck
  • Detection and preliminary physico-chemical properties of antimicrobial components in the native excretions/secretions of three species of Chrysomya (Diptera, Calliphoridae) in Brazil.
Detection and preliminary physico-chemical properties of antimicrobial components in the native excretions/secretions of three species of Chrysomya (Diptera, Calliphoridae) in Brazil.
Acta tropica (2015-03-31)
Norman Arthur Ratcliffe, Cecilia Stahl Vieira, Paloma Martins Mendonça, Rebecca Leal Caetano, Margareth Maria de Carvalho Queiroz, Eloi Souza Garcia, Cicero Brasileiro Mello, Patricia Azambuja
ZUSAMMENFASSUNG
Antibiotic-resistant bacteria in hospitals and communities increasingly threaten public health in Brazil and the rest of the World. There is an urgent need for additional antimicrobial drugs. Calliphorid blowfly larvae are a rich source of antimicrobial factors but the potential of Neotropical species has been neglected. This preliminary study evaluates the antimicrobial activity of the native excretions/secretions of larvae of three species of Brazilian calliphorids, Chrysomya megacephala, Chrysomya albiceps and Chrysomya putoria. Native excretions/secretions were collected from third instar larvae, sterile filtered and tested for antibacterial activity against Staphylococcus aureus 9518, Escherichia coli K12 4401 and Serratia marcescens 365. Turbidometric assays were made in micro-plates, using an ELISA reader, with readings taken up to 22 h. Bacterial suspensions at the start and end of each experiment were also serially diluted, spread on nutrient agar plates and then colony forming units counted. The physico-chemical characteristics of the native excretions/secretions were also tested by freezing/thawing, boiling, and protease digestion. The native excretions/secretions of larvae from these three Chrysomya species significantly inhibited bacterial growth. Therefore, Brazilian calliphorid flies could potentially provide new classes of antibiotics.