Merck
  • Thermodynamic and ultrasonic properties of ascorbic Acid in aqueous protic ionic liquid solutions.
Thermodynamic and ultrasonic properties of ascorbic Acid in aqueous protic ionic liquid solutions.
PloS one (2015-05-27)
Vickramjeet Singh, Gyanendra Sharma, Ramesh L Gardas
ZUSAMMENFASSUNG
In this work, we report the thermodynamic and ultrasonic properties of ascorbic acid (vitamin C) in water and in presence of newly synthesized ammonium based protic ionic liquid (diethylethanolammonium propionate) as a function of concentration and temperature. Apparent molar volume and apparent molar isentropic compression, which characterize the solvation state of ascorbic acid (AA) in presence of protic ionic liquid (PIL) has been determined from precise density and speed of sound measurements at temperatures (293.15 to 328.15) K with 5 K interval. The strength of molecular interactions prevailing in ternary solutions has been discussed on the basis of infinite dilution partial molar volume and partial molar isentropic compression, corresponding volume of transfer and interaction coefficients. Result has been discussed in terms of solute-solute and solute-solvent interactions occurring between ascorbic acid and PIL in ternary solutions (AA + water + PIL).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-(Diethylamino)ethanol, ≥99.5%