Merck
  • Liquid chromatography-tandem mass spectrometry for the quantification of flurbiprofen in human plasma and its application in a study of bioequivalence.
Liquid chromatography-tandem mass spectrometry for the quantification of flurbiprofen in human plasma and its application in a study of bioequivalence.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-05-20)
Chenghan Mei, Bin Li, Qiangfeng Yin, Jing Jin, Ting Xiong, Wenjuan He, Xiujuan Gao, Rong Xu, Piqi Zhou, Heng Zheng, Hui Chen
ZUSAMMENFASSUNG
A simple, quick and accurate LC-MS/MS method for the quantification of flurbiprofen in human plasma with indomethacin as internal standard (IS) was developed and validated. Samples were treated with methanol to precipitate proteins, then separated on a Ultimate C18 column (5μm, 2.1×50mm) with a gradient elusion process. Mobile phase A was comprised of water and formic acid, mobile phase B was comprised of acetonitrile and formic acid. Multi reaction monitoring (MRM) signals were saved on a negative ionization electrospray mass spectrometer. The calibration curve showed good linearity in the range of 40.00-10000.00μg/L (r(2)=0.998). Intra-day RE was 0.2-2.2%. Inter-day RE was 0.5-3.4%. The samples showed good stability under the study conditions. No significant matrix effect was observed. The established method was then applied to a bioequivalence study of a flurbiprofen axetil formulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Flurbiprofen, cyclooxygenase inhibitor