Merck
  • Synthesis and in vitro antiproliferative evaluation of novel hybrids from 1,3,4-thiadiazole and benzisoselenazolone.
Synthesis and in vitro antiproliferative evaluation of novel hybrids from 1,3,4-thiadiazole and benzisoselenazolone.
Chemical & pharmaceutical bulletin (2015-06-02)
Fen Jing, Xiaoyun Fu, Sha Li, Baolin Li, Jijun Zhao, Xuefeng Wang, Yuming Liu, Baoquan Chen
ZUSAMMENFASSUNG
Novel hybrids from 1,3,4-thiadiazole and benzisoselenazolone were designed, synthesized and evaluated for their in vitro antiproliferative activities by CCK-8 assay against three types of human cancer cell lines, SMMC-7721, MCF-7 and A549 cells. The preliminary bioassay results demonstrated that all tested compounds 4a-p showed potent antiproliferative activities, and some compounds exhibited better effects than positive control ethaselen and 5-fluorouracil (5-FU) against various cancer cell lines. Furthermore, compound 4g showed significant antiproliferative activities against SMMC-7721 cells with an IC50 value of 2.08 µM. Compounds 4b and 4m displayed highly effective biological activities against MCF-7 cells with an IC50 values of 2.03 and 2.06 µM, respectively. Compound 4i exhibited the best inhibitory effect against A549 cells with an IC50 value of 1.03 µM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumbromid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Kaliumbromid, powder, 99.999% trace metals basis
Sigma-Aldrich
Kaliumbromid, powder, 99.95% trace metals basis
Sigma-Aldrich
Kaliumbromid, BioXtra, ≥99.0%