Merck
  • Transrectal Rigid-Hybrid Natural Orifice Translumenal Endoscopic Sigmoidectomy for Diverticular Disease: A Prospective Cohort Study.
Transrectal Rigid-Hybrid Natural Orifice Translumenal Endoscopic Sigmoidectomy for Diverticular Disease: A Prospective Cohort Study.
Journal of the American College of Surgeons (2015-08-19)
Sebastian H Lamm, Andreas Zerz, Anastasios Efeoglou, Daniel C Steinemann
ZUSAMMENFASSUNG
Our goal was to evaluate the feasibility of transrectal rigid hybrid natural orifice translumenal endoscopic surgery (NOTES) sigmoidectomy (trNS) in a series of consecutive prospective patients with diverticular disease. The NOTES for left colectomy offers patients reduced pain and easier recovery. Limited data are available for trNS, which is considered safe for various indications. However, the technique is not standardized, and patients in the reported series are highly selected. Patients scheduled for trNS were entered into a prospective registry on an intention-to-treat basis. The primary endpoint was trNS feasibility, and secondary endpoints were morbidity, pain, length of stay, and inflammatory response. A medial-to-lateral dissection with full mobilization of the splenic flexure and total intracorporeal anastomosis was performed. The rectum was covered with a wound protector for transrectal extraction. Of 95 elective sigmoidectomies, 81% (n = 77) were enrolled for either transvaginal NOTES resection (n = 37) or trNS (n = 40). There was no difference in body mass index or indication between patients undergoing laparoscopic-assisted sigmoidectomy (LAS), transvaginal resection, or trNS, although trNS patients were younger. Mainly because of a mismatch of bulky specimen and narrow pelvis, 17.5% of trNS were converted to LAS. Major morbidity was 10%, including 2 septic complications. During the study, the anastomosis technique was changed from double stapled end-to-end to side-to-end anastomosis. Transrectal rigid hybrid natural orifice translumenal endoscopic sigmoidectomy is feasible and safe in a high proportion of unselected consecutive patients with diverticular disease undergoing elective treatment. Intracorporeal side-to-end anastomosis is the preferred technique, and trNS should be offered for elective sigmoidectomy presupposing advanced laparoscopic experience.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kohlenstoffdioxid, ≥99.8%
Sigma-Aldrich
Kohlenstoff-12C-dioxid, 99.9 atom % 12C
Sigma-Aldrich
Kohlenstoff-12C-dioxid, 99.99 atom % 12C