Merck
  • The properties of mesoporous silica nanoparticles functionalized with different PEG-chain length via the disulfide bond linker and drug release in glutathione medium.
The properties of mesoporous silica nanoparticles functionalized with different PEG-chain length via the disulfide bond linker and drug release in glutathione medium.
Journal of biomaterials science. Polymer edition (2015-11-06)
Zhifei Xie, Huameng Gong, Mingxing Liu, Hongda Zhu, Honghao Sun
ZUSAMMENFASSUNG
In this paper, a novel drug-loaded material (MSNs-SS-PEG) was obtained by grafting the thiol-linked methoxy polyethylene glycol (MeOPEG-SH) onto the thiol-functionalized mesoporous silica nanoparticles (MSNs-SH) via the disulfide bond linker. In our designed experiment, three different chain lengths of PEG (PEG(1000), PEG(5000), and PEG(1000)-PEG(5000)) were used. The silica materials were characterized by Fourier transform infrared spectroscopy (FT-IR), dynamic light scattering, field emission scanning electron microscopy, transmission electron microscopy, nitrogen adsorption-desorption measurements, and X-ray diffraction. The morphology of the MSNs-SS-PEG was spherical with an average diameter of about 150 nm. Due to the covalent modification of hydrophilic MeOPEG, the MSNs-SS-PEG was coated by a thin polymer shell, showing stable and inerratic MCM-41 type mesoporous structure as well as high specific surface areas and large pore volumes. Moreover, the releases of doxorubicin hydrochloride (DOX) from these materials at 10 mM of glutathione were investigated. The PEG functionalization could effectively cap drugs in the mesoporous channels. The release of DOX from the MSNs-SS-PEG(n) revealed redox-responsive characteristic. The obtained results showed that the MSNs-SS-PEG might be promising drug delivery carrier materials, which could play an important role in the development of drug delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Doxorubicin -hydrochlorid, 98.0-102.0% (HPLC)
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acryloylchlorid, ≥97%, contains ~400 ppm phenothiazine as stabilizer
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Doxorubicin -hydrochlorid, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Acryloylchlorid, 97.0%, contains <210 ppm MEHQ as stabilizer
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol