Merck
  • Filled hydrogel particles as a delivery system for n-3 long chain PUFA in low-fat frankfurters: Consequences for product characteristics with special reference to lipid oxidation.
Filled hydrogel particles as a delivery system for n-3 long chain PUFA in low-fat frankfurters: Consequences for product characteristics with special reference to lipid oxidation.
Meat science (2015-08-02)
Lorena Salcedo-Sandoval, Susana Cofrades, Claudia Ruiz-Capillas, Francisco Jiménez-Colmenero
ZUSAMMENFASSUNG
This article examines the suitability of filled hydrogel particles for use as a delivery system for n-3 long chain PUFAs in low-fat frankfurters. Their effects on product characteristics over chilled storage were compared with those of frankfurters containing all-pork fat (control) or a comparable amount of fish oil (n-3 LCPUFA) incorporated in liquid form or in an oil-in-water emulsion. In modified samples n-3 fatty acids ranged between 801.34 to 996.37 mg/100g as opposed to 66 mg/100g in all-pork fat product. As compared with the control, hardness and chewiness values were similar (P>0.05) in filled hydrogel frankfurter. The presence of fish oil favoured lipid oxidation to varying degrees depending on delivery system, in descending order: direct oil addition>oil-in-water emulsion>hydrogels. Sensory evaluation demonstrated the advantages, from a sensory point of view, of hydrogel filled particles as n-3 delivery systems in frankfurters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Cumolhydroperoxid, technical grade, 80%
Sigma-Aldrich
2-Thiobarbitursäure, ≥98%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
1,1,3,3-Tetraethoxypropan, ≥96%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.