Merck
  • Microdiplanol and microdiplane: a new m-anisaldehyde and a new 24-methylcholestanol derivative from the endophytic fungus Microdiplodia sp.
Microdiplanol and microdiplane: a new m-anisaldehyde and a new 24-methylcholestanol derivative from the endophytic fungus Microdiplodia sp.
Journal of Asian natural products research (2015-01-07)
Hidayat Hussain, Karsten Krohn, Ahmed Al-Harrasi, Simeon F Kouam, Gulam Abbas, Muhammad Adil Raees, Thomas Dzeha, Riaz Ullah, Aqib Zahoor, Afzal Shah, Amin Badshah, Amjad Khan, Iftikhar Ali, Barbara Schulz
ZUSAMMENFASSUNG
Phytochemical investigation of the endophytic fungus Microdiplodia sp. afforded a new m-anisaldehyde derivative named microdiplanol (1) and a new 24-methylcholestanol derivative named microdiplane (2). Their structures were confirmed by a comprehensive analysis of 1D and 2D NMR and mass spectrometric data.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cer(IV)-sulfat