Merck
The role of glucocorticoid receptor phosphorylation in the model of negative affective states.
The world journal of biological psychiatry : the official journal of the World Federation of Societies of Biological Psychiatry (2015-03-10)
Milica Jovicic, Nadja P Maric, Ivan Soldatovic, Iva Lukic, Sanja Andric, Marina Mihaljevic, Zorana Pavlovic, Milos Mitic, Miroslav Adzic
ZUSAMMENFASSUNG
To develop a structural equation model of negative affectivity (NA) that involves interaction of glucocorticoid receptor (GR) signaling, personality dimensions and recent stressful life events. Seventy participants - 35 diagnosed with major depression and 35 healthy controls, were enrolled in the study. Morning plasma cortisol levels were determined by chemiluminescent immunometric assays. Molecular parameters (total nuclear and cytoplasmatic GR, nuclear GR phosphorylated at serine 211 (pGR-211) and at serine 226 (pGR-226) and cytoplasmic FKBP51) were analysed from peripheral blood lymphocytes by Western blot. NA, personality dimensions and stressful life events were assessed by self-report instruments. GR signalling parameters had direct independent effect on measures of NA, with pGR-226 levels showing the strongest correlation, followed by FKBP51 and pGR-211 levels. Neuroticism and extraversion also demonstrated strong independent effect on NA, while recent stressful events did not predict NA directly, but demonstrated a significant effect on personality dimensions. Cortisol, total nuclear GR and total cytoplasmatic GR levels were excluded from the model due to non-significant correlations with NA. Negative affectivity is a transdiagnostic factor in vulnerability to affective disorders and possible therapeutic target. Molecular signature of negative affectivity should incorporate GR phosphorylation with other known biological underpinnings.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri