Merck
  • (18) F-FDG PET/CT for initial staging in breast cancer patients - Is there a relevant impact on treatment planning compared to conventional staging modalities?
(18) F-FDG PET/CT for initial staging in breast cancer patients - Is there a relevant impact on treatment planning compared to conventional staging modalities?
European radiology (2015-02-15)
J Krammer, A Schnitzer, C G Kaiser, K A Buesing, E Sperk, J Brade, S Wasgindt, M Suetterlin, S O Schoenberg, E J Sutton, K Wasser
ZUSAMMENFASSUNG
To evaluate the impact of whole-body (18) F-FDG PET/CT on initial staging of breast cancer in comparison to conventional staging modalities. This study included 102 breast cancer patients, 101 patients were eligible for evaluation. Preoperative whole-body staging with PET/CT was performed in patients with clinical stage ≥ T2 tumours or positive local lymph nodes (n = 91). Postoperative PET/CT was performed in patients without these criteria but positive sentinel lymph node biopsy (n = 10). All patients underwent PET/CT and a conventional staging algorithm, which included bone scan, chest X-ray and abdominal ultrasound. PET/CT findings were compared to conventional staging and the impact on therapeutic management was evaluated. PET/CT led to an upgrade of the N or M stage in overall 19 patients (19 %) and newly identified manifestation of breast cancer in two patients (2 %). PET/CT findings caused a change in treatment of 11 patients (11 %). This is within the range of recent studies, all applying conventional inclusion criteria based on the initial T and N status. PET/CT has a relevant impact on initial staging and treatment of breast cancer when compared to conventional modalities. Further studies should assess inclusion criteria beyond the conventional T and N status, e.g. tumour grading and receptor status. • PET/CT may be relevant in staging breast cancer patients at higher risk for metastases • PET/CT may modify the N and M stage in multiple patients • PET/CT may impact treatment planning in breast cancer patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tantal(V)-ethoxid, 99.98% trace metals basis
Tantal(V)-ethoxid, packaged for use in deposition systems