Merck
  • EFFECT OF PREGNANE XENOBIOTIC RECEPTOR ACTIVATION ON INFLAMMATORY BOWEL DISEASE TREATED WITH RIFAXIMIN.
EFFECT OF PREGNANE XENOBIOTIC RECEPTOR ACTIVATION ON INFLAMMATORY BOWEL DISEASE TREATED WITH RIFAXIMIN.
Journal of biological regulators and homeostatic agents (2015-07-01)
Y C Wan, T Li, Y-D Han, H-Y Zhang, H Lin, B Zhang
ZUSAMMENFASSUNG
The causes and pathogenesis of Inflammatory Bowel Disease (IBD) are still not clearly understood. This study aims to prove the important role of rifaximin played in inflammatory reaction caused by abnormity of the intestinal mucosal immune system. Intestinal microflora can greatly promote and maintain the inflammatory reaction of IBD, therefore, antibiotics can be used to treat IBD. Rifaximin is a medicine usually used for local intestinal infection. Many clinical and basic studies have shown that both a single application of rifaximin and the joint application with other medicines could achieve a good efficacy. This paper studied the activation of Pregnane Xenobiotic Receptor (PXR) in treating IBD with rifaximin and analyzed its efficacy in IBD when PXR was involved in the transport of medicine and metabolism. The results prove that rifaximin can not only serve as an anti-microbial drug, but can activate PXR and actually weaken the reaction of IBD. Thus it is safe to say that rifaximin has great potential in treating IBD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals