Merck
  • Luteal-long GnRH agonist versus flexible-multidose GnRH antagonist protocols for overweight and obese patients who underwent ICSI.
Luteal-long GnRH agonist versus flexible-multidose GnRH antagonist protocols for overweight and obese patients who underwent ICSI.
Journal of obstetrics and gynaecology : the journal of the Institute of Obstetrics and Gynaecology (2014-09-23)
I Esinler, G Bozdag, D Esinler, K S Lale, H Yarali
ZUSAMMENFASSUNG
A total of 413 consecutive infertile patients (572 cycles) with a body mass index (BMI) of ≥ 25 kg/m(2) were enrolled into the study. The luteal-long GnRH agonist group (Group I) constituted 211 patients (300 cycles) and the flexible-multidose GnRH antagonist group (Group II) constituted 202 patients (272 cycles). The duration of stimulation (d) (10.1 ± 2.5 vs. 9.2 ± 2.0; p < 0.01); the total dose of gonadotrophin used (IU) (3,099.4 ± 2,885.0 vs. 2,684.0 ± 1,046.4; p < 0.05) and the E2 level on the day of hCG (pg/ml) (2,375.8 ± 1,554.6 vs. 1,905.6 ± 1,598.8; p < 0.01) were significantly lower in Group II when compared with Group I. However, the ongoing pregnancy per embryo transfer (37.0% vs. 25.7%; p < 0.05) and the implantation rate (25.7% vs. 15.6%; p < 0.01) were significantly lower in Group II when compared with Group I. In conclusion, we noted that the luteal-long GnRH agonist protocol produced higher implantation rates and higher clinical-ongoing pregnancy rates in overweight and obese patients when compared with the flexible-multidose GnRH antagonist protocol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture