Merck
  • Candida rugosa lipase immobilization on hydrophilic charged gold nanoparticles as promising biocatalysts: Activity and stability investigations.
Candida rugosa lipase immobilization on hydrophilic charged gold nanoparticles as promising biocatalysts: Activity and stability investigations.
Colloids and surfaces. B, Biointerfaces (2015-05-15)
Iole Venditti, Cleofe Palocci, Laura Chronopoulou, Ilaria Fratoddi, Laura Fontana, Marco Diociaiuti, Maria Vittoria Russo
ZUSAMMENFASSUNG
In this work, a simple and versatile methodology to obtain two different bioconjugated systems has been developed by the immobilization of Candida rugosa lipase (CRL) on hydrophilic gold nanoparticles functionalized with 2-diethylaminoethanethiol hydrochloride (DEA) or with sodium 3-mercapto-1-propanesulfonate (3MPS), namely Au-DEA@CRL and Au-3MPS@CRL. Both spectroscopic and morphological properties of metal nanoparticles have been deeply investigated. The enzyme loading and lipolytic activity of AuNPs@CRL bioconjugates have been studied with respect to different surface functionalization and compared with the free enzyme. Some physical and chemical parameters had a strong effect on enzyme activity and stability, that were improved in the case of the Au-DEA@CRL bioconjugate, which showed a remarkable biocatalytic performance (95% of residual lipolytic activity compared with free CRL) and stability in experimental conditions concerning pH (range 5-8) and temperature (range 20-60°C), as often required for the industrial scale up of catalytic systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Natriumborhydrid, powder, ≥98.0%
Sigma-Aldrich
Natriumborhydrid, ReagentPlus®, 99%
Sigma-Aldrich
Natriumborhydrid, granular, 99.99% trace metals basis
Sigma-Aldrich
Phenolphthalein, ACS reagent
Sigma-Aldrich
Glycerin-tributyrat, ≥99%
Sigma-Aldrich
Natriumborhydrid, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Phenolphthalein -Lösung, 0.5 wt. % in ethanol: water (1:1)
Sigma-Aldrich
Natriumborhydrid, granular, 10-40 mesh, 98%
Sigma-Aldrich
O-Acetyl-L-Carnitin -hydrochlorid, ≥99% (titration), powder
Sigma-Aldrich
Natriumborhydrid -Lösung, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Natriumborhydrid, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Natriumborhydrid -Lösung, 2.0 M in triethylene glycol dimethyl ether