Merck
  • Preventive effect of Pueraria mirifica on testosterone-induced prostatic hyperplasia in Sprague Dawley rats.
Preventive effect of Pueraria mirifica on testosterone-induced prostatic hyperplasia in Sprague Dawley rats.
Andrologia (2015-01-21)
S S Masrudin, J Mohamad
ZUSAMMENFASSUNG
Pueraria mirifica (PM) extract contains phytoestrogen daidzein and genistein. In this study, we investigated the protective effect of PM extract, daidzein and genistein on a testosterone-induced prostatic hyperplasia in rats. Testosterone was administered at 3 mg kg(-1) to rats followed by the PM extract, daidzein and genistein for a period of 30 days with finasteride as positive control. The testosterone level was increased, indicating inhibition of 5α-reductase converting testosterone to dihydrotestosterone. This was confirmed by prostate-specific antigen level that significantly decreased when treated with PM extract, daidzein and genistein. The PM extract, daidzein and genistein reduced the increase in the prostate/body weight ratio in testosterone-induced rats. This gives indication that PM extract, daidzein and genistein possessed protective activity for the treatment of benign prostatic hyperplasia. The analysis of histoarchitechture of the prostate has also shown that there was a significant improvement in prostatic cells of the testosterone-induced rats when treated with PM extract, daidzein and genistein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Testosteron, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Finasterid, ≥98% (HPLC), powder
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)