Merck
  • Drug delivery with nanospherical supramolecular cell penetrating peptide-taxol conjugates containing a high drug loading.
Drug delivery with nanospherical supramolecular cell penetrating peptide-taxol conjugates containing a high drug loading.
Journal of colloid and interface science (2015-05-10)
Ran Tian, Huaimin Wang, Ruifang Niu, Dan Ding
ZUSAMMENFASSUNG
Supramolecular nanostructures via small molecule self-assembly hold great promise for controlled delivery of hydrophobic anticancer drugs. Particularly, taxol has recently been discovered to possess excellent self-assembly property, which may provide new opportunities to develop a new class of functional supramolecular nanomaterials for drug delivery application. A cell penetrating peptide (CPP)-taxol conjugate (Taxol-CPP) was designed and synthesized. The self-assembling property of Taxol-CPP was investigated and the resultant nanomaterials were well characterized. Subsequently, the cytotoxicity of the Taxol-CPP after self-assembly against HepG2 cancer cells was evaluated. It is found that the Taxol-CPP possesses a high drug loading of 26.4% in each molecule, which is able to self-assemble into supramolecular nanospheres. By taking advantages of the self-assembly ability of taxol, Taxol-CPP supramolecular nanospheres with a mean size of around 130 nm can be obtained, composed of only the functional peptide (CPP) and the drug (taxol). Furthermore, we have demonstrated that the Taxol-CPP nanospheres do not compromise the taxol's potency, which can also be utilized as the carriers for co-delivery of another anticancer drug (doxorubicin).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Bernsteinsäureanhydrid, ≥99% (GC)
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder