Merck
  • Chelation-assisted substrate-controlled asymmetric lithiation-allylboration of chiral carbamate 1,2,4-butanetriol acetonide.
Chelation-assisted substrate-controlled asymmetric lithiation-allylboration of chiral carbamate 1,2,4-butanetriol acetonide.
Molecules (Basel, Switzerland) (2015-06-02)
Adeem Mahmood, Hamad Z Alkhathlan, Saima Parvez, Merajuddin Khan, Sohail A Shahzad
ZUSAMMENFASSUNG
The lithiation of 2-(2,2-dimethyl-1,3-dioxolan-4-yl)ethyl diisopropylcarbamate (1) is achieved freely by sec-butyllithium in diethylether with high lk-diastereoselectivity: the bicyclic chelate complexes 3a and 3b are reacted with electrophiles to form optically active precursors 4a and 4b with >95% diastereoselectivity. In addition, tertiary diamines can undergo an external complexation in contest with the internal oxygen ligand, leading to improved stereoselectivities. The further reactions of lithiated carbamates with trans alkenyl-9-BBN derivatives after 1,2 metallate rearrangements, gave the key intermediate α-substituted allylic boranes 7. Subsequent allylboration of aldehydes gave (Z)-anti-homoallylic alcohols 8 in good yield and excellent d.r.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
sec-Butyllithium -Lösung, 1.4 M in cyclohexane
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
Cyclohexan, anhydrous, 99.5%