Merck
  • The dual targeting of EGFR and ErbB2 with the inhibitor Lapatinib corrects high glucose-induced apoptosis and vascular dysfunction by opposing multiple diabetes-induced signaling changes.
The dual targeting of EGFR and ErbB2 with the inhibitor Lapatinib corrects high glucose-induced apoptosis and vascular dysfunction by opposing multiple diabetes-induced signaling changes.
Journal of drug targeting (2015-06-27)
Ibrahim F Benter, Fatima Sarkhou, Abeer T Al-Khaldi, Bindu Chandrasekhar, Sreeja Attur, Gursev S Dhaunsi, Mariam H M Yousif, Saghir Akhtar
ZUSAMMENFASSUNG
The epidermal growth factor receptors, EGFR and EGFR2 (ErbB2), appear important mediators of diabetes-induced vascular dysfunction. We investigated whether targeted dual inhibition of EGFR and ErbB2 with Lapatinib would be effective in treating diabetes-induced vascular dysfunction in a rat model of type 1 diabetes. In streptozotocin-induced diabetes, chronic 4-week oral or acute, ex vivo, administration of Lapatinib prevented the development of vascular dysfunction as indicated by the attenuation of the hyper-reactivity of the diabetic mesenteric vascular bed (MVB) to norephinephrine without correcting hyperglycemia. Chronic in vivo or acute ex vivo Lapatinib treatment also significantly attenuated diabetes-induced increases in phosphorylation of EGFR, ErbB2, ERK1/2, AKT, ROCK2 and IkB-alpha as well as normalized the reduced levels of phosphorylated FOXO3A, and eNOS (Ser1177) in the diabetic MVB. Similar results were observed in vascular smooth muscle cells (VSMCs) cultured in high glucose (25 mM) treated with Lapatinib or small interfering RNA (siRNA) targeting the ErbB2 receptor. Lapatinib also prevented high glucose-induced apoptosis in VSMC. Thus, Lapatinib corrects hyperglycemia-induced apoptosis and vascular dysfunction with concomitant reversal of diabetes or high glucose-induced signaling changes in EGFR/ErbB2 and downstream signaling pathways implying that targeted dual inhibition of EGFR/ErbB2 might be an effective vasculoprotective treatment strategy in diabetic patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Acrylnitril, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Acrylnitril, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri