Merck
  • Morphological Changes in the Overall Mycobacterium tuberculosis H37Ra Cell Shape and Cytoplasm Homogeneity due to Mutellina purpurea L. Essential Oil and Its Main Constituents.
Morphological Changes in the Overall Mycobacterium tuberculosis H37Ra Cell Shape and Cytoplasm Homogeneity due to Mutellina purpurea L. Essential Oil and Its Main Constituents.
Medical principles and practice : international journal of the Kuwait University, Health Science Centre (2015-09-19)
Elwira Sieniawska, Marta Swatko-Ossor, Rafał Sawicki, Grażyna Ginalska
ZUSAMMENFASSUNG
The aim of this study was to evaluate the antimycobacterial activity of the essential oil (EO) of Mutellina purpurea L. and its main constituents against the M. tuberculosis H37Ra strain. The M. purpurea EO was obtained by hydrodistillation, while its main constituents were purchased. The minimal inhibitory concentration values were determined by the log2 dilution method. Visualization of the effects of the tested substances on M. tuberculosis was performed using a transmission electron microscope (TEM). Mathematical shape descriptors such as area, circularity, aspect ratio and roundness were calculated to describe morphological changes in bacterial cell shape. The EO of M.purpurea and all substances tested in this experiment showed a significant antimycobacterial activity. The most active was α-pinene followed by bisabolol and myrcene (8, 16 and 32 µg/ml, respectively). The EO and limonene exhibited the same antimicrobial activity (64 µg/ml). The TEM images and shape descriptors showed significant changes in the overall tuberculosis cell shape and cytoplasm homogeneity (uniformity and consistency) In this study, the low molecular weight compounds of mono- and sesquiterpenes penetrated/destabilized the complex mycobacterial cell wall and decreased its viability. There is a need for further experiments to explain the mechanism of action of these small particles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Supelco
D-(+)-Glukose, analytical standard
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Myrcen, technical grade
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Myrcen, stabilized, FCC, FG
Sigma-Aldrich
D-(+)-Glukose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glukose, 99.9 atom % 16O, 99.9 atom % 12C