Merck
  • Encapsulation of sulfur with thin-layered nickel-based hydroxides for long-cyclic lithium-sulfur cells.
Encapsulation of sulfur with thin-layered nickel-based hydroxides for long-cyclic lithium-sulfur cells.
Nature communications (2015-10-17)
Jian Jiang, Jianhui Zhu, Wei Ai, Xiuli Wang, Yanlong Wang, Chenji Zou, Wei Huang, Ting Yu
ZUSAMMENFASSUNG
Elemental sulfur cathodes for lithium/sulfur cells are still in the stage of intensive research due to their unsatisfactory capacity retention and cyclability. The undesired capacity degradation upon cycling originates from gradual diffusion of lithium polysulfides out of the cathode region. To prevent losses of certain intermediate soluble species and extend lifespan of cells, the effective encapsulation of sulfur plays a critical role. Here we report an applicable way, by using thin-layered nickel-based hydroxide as a feasible and effective encapsulation material. In addition to being a durable physical barrier, such hydroxide thin films can irreversibly react with lithium to generate protective layers that combine good ionic permeability and abundant functional polar/hydrophilic groups, leading to drastic improvements in cell behaviours (almost 100% coulombic efficiency and negligible capacity decay within total 500 cycles). Our present encapsulation strategy and understanding of hydroxide working mechanisms may advance progress on the development of lithium/sulfur cells for practical use.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, ACS reagent, ≥99.0%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
1-Methyl-2-pyrrolidon, ReagentPlus®, 99%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, anhydrous, 99.5%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
1-Methyl-2-pyrrolidon, biotech. grade, ≥99.7%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hexamethylentetramin, ACS reagent, ≥99.0%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hexamethylentetramin, ReagentPlus®, 99%
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hexamethylentetramin, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Sigma-Aldrich
Hexamethylentetramin, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)