Merck
  • Increased circulating heat shock protein 70 (HSPA1A) levels in gestational diabetes mellitus: a pilot study.
Increased circulating heat shock protein 70 (HSPA1A) levels in gestational diabetes mellitus: a pilot study.
Cell stress & chaperones (2015-02-28)
Zoltán Garamvölgyi, Zoltán Prohászka, János Rigó, András Kecskeméti, Attila Molvarec
ZUSAMMENFASSUNG
Recent data indicate that serum Hsp70 (HSPA1A) levels are increased in type 1 and 2 diabetes mellitus. However, there is no report in the literature on circulating Hsp70 levels in gestational diabetes mellitus. In this pilot study, we measured serum Hsp70 levels in 11 pregnant women with pregestational diabetes, 38 women with gestational diabetes, and 40 healthy pregnant women with ELISA. Plasma glucose levels, serum insulin concentrations, HbA1c values, and the Homeostatic Model Assessment-Insulin Resistance (HOMA-IR) index were also determined. According to our results, serum Hsp70 concentrations were significantly higher in women with pregestational and gestational diabetes mellitus than in healthy pregnant women. In addition, pregestational diabetic women had significantly higher Hsp70 levels than those with gestational diabetes. Furthermore, in the group of women with gestational diabetes mellitus, serum Hsp70 levels showed a significant positive correlation with HbA1c values. However, there was no other relationship between clinical features and metabolic parameters of the study subjects and their serum Hsp70 levels in either study group. In conclusion, we demonstrated for the first time in the literature that serum Hsp70 levels are increased and correlate with HbA1c values in women with gestational diabetes mellitus. Nevertheless, further studies are needed to determine whether circulating Hsp70 plays a causative role in the pathogenesis of gestational diabetes or elevated serum Hsp70 levels are only consequences of the disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
o-Phenylendiamin, flaked, 99.5%
Sigma-Aldrich
o-Phenylendiamin, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylendiamin, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylendiamin, sublimed, ≥99%