Merck
  • Preparation of polyclonal antibodies for nateglinide (NTG) and development of a sensitive chemiluminescent immunoassay to detect NTG in tablets and serum.
Preparation of polyclonal antibodies for nateglinide (NTG) and development of a sensitive chemiluminescent immunoassay to detect NTG in tablets and serum.
Talanta (2015-12-24)
Lei Zheng, Jing Wang, Jie Zhang, Zhaorui Song, Yaqing Dong, Yufen Wang, Zhongsheng Tong, Chuan Deng, Yongmei Yin, Meng Meng, Rimo Xi
ZUSAMMENFASSUNG
In this study, we prepared polyclonal antibodies against anti-diabetic drug nateglinide (NTG), and established a sensitive chemiluminescent immunoassay (CLIA) to detect NTG in tablets and serum. Two kinds of immunogens were synthesized using ethylcarbodiimide (EDC)/hydroxysuccinimide (NHS) and carbonyldiimidazole (CDI)/4-dimethylaminopyridine (DMAP) as coupling reagents respectively. When activated by EDC/NHS, more molecules of NTG coupled with carrier protein in immunogens. A horseradish peroxidase (HRP)-luminol-H2O2 system with p-iodophenol enhancement was applied in the CLIA analysis. The antibodies in EDC/NHS group showed higher titer, sensitivity and wider detection linear range than those in CDI/DMAP group, and were chosen for next studies. The developed CLIA assay exhibited good selectivity towards NTG among structually similar analogs. The method could detect as low as 0.35 ng mL(-1) NTG in buffer, 2.1 ng mL(-1) NTG in serum and 0.84 ng mL(-1) NTG in tablets. The CLIA method provided consistent results with HPLC method (r=0.9986) in determination of NTG from 5.0 to 400 µg mL(-1). The CLIA method could detect 78 samples in one assay, and the samples need only dilution in pretreatment. As a summary, this research offers a sensitive assay for high-throughout screening of NTG in formulation control and pharmacokinetic studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Picrylsulfonsäure -Lösung, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
4-(Dimethylamino)-pyridin, ReagentPlus®, ≥99%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥99%
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
1,1′-Carbonyldiimidazol, reagent grade
Sigma-Aldrich
1,1′-Carbonyldiimidazol, ≥97.0% (T)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98% (TLC)
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98.0% (NT)
Sigma-Aldrich
4-Jodphenol, 99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, tablet, 1 mg substrate per tablet