Merck
  • Overexpression of the laeA gene leads to increased production of cyclopiazonic acid in Aspergillus fumisynnematus.
Overexpression of the laeA gene leads to increased production of cyclopiazonic acid in Aspergillus fumisynnematus.
Fungal biology (2015-10-16)
Eun Jin Hong, Na Kyeong Kim, Doyup Lee, Won Gon Kim, Inhyung Lee
ZUSAMMENFASSUNG
To explore novel bioactive compounds produced via activation of secondary metabolite (SM) gene clusters, we overexpressed an ortholog of laeA, a gene that encodes a global positive regulator of secondary metabolism in Aspergillus fumisynnematus F746. Overexpression of the laeA gene under the alcA promoter resulted in the production of less pigment, shorter conidial head chains, and fewer conidia. Thin-layer chromatography (TLC) and high-performance liquid chromatography (HPLC) analysis revealed that SM production in OE::laeA was significantly increased, and included new metabolites that were not detected in the wild type. Among them, a compound named F1 was selected on the basis of its high production levels and antibacterial effects. F1 was purified by column chromatography and preparative TLC and identified as cyclopiazonic acid (CPA) by LC/MS, which had been previously known as mycotoxin. As A. fumisynnematus was not known to produce CPA, these results suggest that overexpression of the laeA gene can be used to explore the synthesis of useful bioactive compounds, even in a fungus for which the genome sequence is unavailable.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
2,3,5-Triphenyltetrazoliumchlorid, ≥98.0% (HPLC)
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Cyclopentanon, ≥99%, FG
Sigma-Aldrich
Cyclopentanon, ReagentPlus®, ≥99%
Sigma-Aldrich
N6-Cyclopentyladenosine, solid