Merck
Cholesterol metabolites exported from human brain.
Steroids (2015-02-11)
Luigi Iuliano, Peter J Crick, Chiara Zerbinati, Luigi Tritapepe, Jonas Abdel-Khalik, Marc Poirot, Yuqin Wang, William J Griffiths
ZUSAMMENFASSUNG
The human brain contains approximately 25% of the body's cholesterol. The brain is separated from the circulation by the blood brain barrier. While cholesterol will not passes this barrier, oxygenated forms of cholesterol can cross the barrier. Here by measuring the difference in the oxysterol content of blood plasma in the jugular vein and in a forearm vein by mass spectrometry (MS) we were able to determine the flux of more than 20 cholesterol metabolites between brain and the circulation. We confirm that 24S-hydroxycholesterol is exported from brain at a rate of about 2-3mg/24h. Gas chromatography (GC)-MS data shows that the cholesterol metabolites 5α-hydroxy-6-oxocholesterol (3β,5α-dihydroxycholestan-6-one), 7β-hydroxycholesterol and 7-oxocholesterol, generally considered to be formed through reactive oxygen species, are similarly exported from brain at rates of about 0.1, 2 and 2mg/24h, respectively. Although not to statistical significance both GC-MS and liquid chromatography (LC)-MS methods indicate that (25R)26-hydroxycholesterol is imported to brain, while LC-MS indicates that 7α-hydroxy-3-oxocholest-4-enoic acid is exported from brain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Hexamethyldisilazan, reagent grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Hexamethyldisilazan, ReagentPlus®, 99.9%
Avanti
3β,7α-Dihydroxycholest-5-enoic acid-d3, Avanti Research - A Croda Brand, powder