Merck
  • Pure total flavonoids from Citrus paradisi Macfadyen act synergistically with arsenic trioxide in inducing apoptosis of Kasumi-1 leukemia cells in vitro.
Pure total flavonoids from Citrus paradisi Macfadyen act synergistically with arsenic trioxide in inducing apoptosis of Kasumi-1 leukemia cells in vitro.
Journal of Zhejiang University. Science. B (2015-07-15)
Bo Wang, Sheng-yun Lin, Ying-ying Shen, Li-qiang Wu, Zhen-zhen Chen, Jing Li, Zhi Chen, Wen-bin Qian, Jian-ping Jiang
ZUSAMMENFASSUNG
To investigate the potential effects of pure total flavonoid compounds (PTFCs) from Citrus paradisi Macfadyen separately or combined with arsenic trioxide on the proliferation of human myeloid leukemia cells and the mechanisms underlying the action of PTFCs. The effects of PTFCs separately or combined with arsenic trioxide on the proliferation and apoptosis of leukemia cells were determined by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT), fluorescence microscopy, and flow cytometry. Their effects on the expression levels of apoptosis-related regulators were determined by Western blot assay. PTFCs combined with arsenic trioxide significantly inhibited the growth of Kasumi-1 cells, and apoptosis was confirmed by flow cytometry analysis. Hoechst 33258 staining showed more significant morphological changes and more apoptosis following the combined treatment. Western blots showed changes in the expression of genes for poly ADP-ribose polymerase (PARP), caspase 3/9, and P65. The results indicated that PTFCs separately or combined with arsenic trioxide inhibited proliferation of leukemia cells in vitro and induced their apoptosis by modulating the expression of apoptosis-related regulator genes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)