Merck
  • Application of hollow fiber liquid phase microextraction for simultaneous determination of regulated and emerging iodinated trihalomethanes in drinking water.
Application of hollow fiber liquid phase microextraction for simultaneous determination of regulated and emerging iodinated trihalomethanes in drinking water.
Journal of chromatography. A (2015-06-01)
A Domínguez-Tello, A Arias-Borrego, T García-Barrera, J L Gómez-Ariza
ZUSAMMENFASSUNG
Trihalomethanes (THMs) are regulated disinfection by-products (DBPs) most commonly analyzed in quality control water supply due to their harmful effects on health. However, few data exist about the content of emerging iodo-trihalomethanes (I-THMs) which are present in drinking water at very low concentrations (in the order of ngL(-1)). For this reason a two-phase hollow fiber liquid phase microextraction method for the simultaneous determination of four regulated trihalomethanes and six emerging iodo-trihalomethanes using GC-μECD and GC-MS with detection limits in the range of few ngL(-1) has been developed. A central composite design was used to optimize conditions for simultaneous extraction. The best extraction recovery was obtained with 19.2min at 27.1°C and 900rpm, without salt addition, using a supported hollow fiber membrane of 10.5cm (0.6mm id) and 1-octanol as acceptor phase. The limits of detection for the regulated THMs and I-THMs were 3-44ngL(-1) and 1-3ngL(-1), respectively. The calibration curves showed good linearity (R(2)>0.995) and good repeatibility (3-22%). The relative recoveries in water were between 96.5% and 105.2%. The method was applied for the simultaneous determination of trihalomethanes in supply water samples from seven water distribution systems (WDS) in the Huelva area, located at the southwest Spain, which use different water-treatment processes. The highest concentrations of I-THMs, particularly CHBrClI and CHCl2I, were detected in water treated with advanced treatment process using pre-ozonation, however these compounds were not detected or decreased along distribution system. In the samples of treated water with conventional treatment, using pre-oxidation by permanganate and distribution network, CHCl2I, CHBrClI, CHClI2, CHBrI2 and CHI3 were detected at very low concentrations (1-18ngL(-1)). Finally, in water samples from underground origin without oxidation treatment, in which only disinfection with sodium hypochlorite was applied, I-THMs were not detected.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Bromoform, contains 60-120 ppm 2-methyl-2-butene as stabilizer, 99%
Sigma-Aldrich
Iodoform, 99%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Bromoform, contains 1-3% ethanol as stabilizer, 96%
Sigma-Aldrich
Dibromchlormethan, 97%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC