Merck
Surveillance of perchlorate in ground water, surface water and bottled water in Kerala, India.
Journal of environmental health science & engineering (2015-07-30)
Anupama Vijaya Nadaraja, Prajeesh Gangadharan Puthiyaveettil, Krishnakumar Bhaskaran
ZUSAMMENFASSUNG
Perchlorate is an emerging water contaminant that disrupts normal functioning of human thyroid gland and poses serious threat to health, especially for pregnant women, fetus and children. High level of perchlorate contamination in fresh water sources at places nearby ammonium perchlorate (rocket fuel) handled in bulk is reported in this study. Of 160 ground water samples analyzed from 27 locations in the State Kerala, 58 % had perchlorate above detection limit (2 μg/L) and the highest concentration observed was 7270 μg/L at Ernakulam district, this value is ~480 times higher than USEPA drinking water equivalent level (15 μg/L). Perchlorate was detected in all surface water samples analyzed (n = 10) and the highest value observed was 355 μg/L in Periyar river (a major river in the State). The bottled drinking water (n = 5) tested were free of perchlorate. The present study underlines the need for frequent screening of water sources for perchlorate contamination around places the chemical is handled in bulk. It will help to avoid human exposure to high levels of perchlorate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O